Learn More
Hettich Lab Technology™ MIKRO 220 Microlitre Centrifuge
Click to view available options
Capacity:
48 x 1.5/2.0 mL
Unit Size:
Each
Description
- Metal casing and lid
- Centrifugation chamber of rust-free stainless steel
- View port in the lid
- 9 different rotors: In addition to the standard rotors, it has a 24-place swing-out rotor, a 20-place rotor for cryotubes and a rotor that can hold 6 PCR strips
- Motor-driven lid locking
- Ergonomic operating and information display
- Simple rotor change
- 10 programmable memories
- Entry of the rotor radius in mm enables switching between display of rpm and RCF (relative centrifugal force)
- Parameters can be entered or changed rapidly and easily using the rotary button
- Lid locked when rotor is running
- Lid fall prevention mechanism
- Emergency lid release in the event of power failure
- Motor overheating protection
- Imbalance shut-off
- Automatic rotor recognition
- Accessories with bioseal (TÜV tested in accordance with DIN EN 61010, Part 2) to protect against dangerous or aggressive aerosols
Specifications
Specifications
| Capacity | 48 x 1.5/2.0 mL |
| Max. Speed | 18,000 rpm |
| Max. RCF | 31,514 x g |
| Height (English) Exterior | 12.3 in. |
| Height (Metric) Exterior | 313 mm |
| Width (English) Exterior | 13 in. |
| Width (Metric) Exterior | 330 mm |
| Type | Microlitre Centrifuge |
| Depth (English) | 16.5 in. |
| Depth (Metric) | 420 mm |
| Show More |