missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIDN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18939-1-AP-150UL
This item is not returnable.
View return policy
Description
Expression of the midnolin gene is developmentally regulated: it is strongly expressed at the mesencephalon (midbrain) of the embryo in day 12.5 (E12.5) mice. The midnolin encodes a protein of 508 amino acids (aa), which contains a Ubiquitin-like domain. The intracellular distribution of the midnolin was studied by using midnolin-green fluorescent protein (GFP) fusion proteins. Midnolin was found to be localized in the nucleus and nucleolus, but not in the cytoplasm. The nucleolar localization signal was determined to be a 28aa peptide located at the C-terminal region of the midnolin. May be involved in regulation of genes related to neurogenesis in the nucleolus.Specifications
| MIDN | |
| Polyclonal | |
| Unconjugated | |
| MIDN | |
| Midbrain nucleolar protein, MIDN, midnolin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 314623, 59090, 90007 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D4AE48, Q3TPJ7, Q504T8 | |
| Midn | |
| MIDN Fusion Protein Ag13492 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction