Learn More
IKA™ Midi MR 1 Digital Magnetic Stirrer
Product Code. 15670450 Shop All IKA Products
Change view
Click to view available options
Capacity:
50 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15670450 50 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15670450 Supplier IKA™ Supplier No. 0025002968
  • ONLINE EXCLUSIVE PRICE
    56320.00 NOK
    49800.07 NOK / Pack of 1
    Save 6519.93 NOK
    11% Off
View all promotions
Estimated Shipment: 13-11-2026
Powerful magnetic stirrer without heating.
  • Flat, sturdy stainless steel casing
  • Non-locking motor
  • Infinitely variable speed
  • Timer or continuous operation
  • Integrated USB and RS 232 interface
TRUSTED_SUSTAINABILITY
Specifications
Speed 0 to 1000 rpm
Top Plate Material Stainless Steel
Capacity 50 L
Dimensions (L x W x H) 360 x 110 x 430 mm
Electrical Requirements 230/100 - 120/100 V, 50/60 Hz, 70 W
Includes RS 232, USB Interface
Motor Type Non-locking
Weight (Metric) 10.7 kg