missing translation for 'onlineSavingsMsg'
Learn More

MID1IP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16855084 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16855084 20 μL 20µL
16845084 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16855084

Brand: Proteintech 157641AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Plays a role in the regulation of lipogenesis in liver. Up-regulates ACACA enzyme activity. Required for efficient lipid biosynthesis, including triacylglycerol, diacylglycerol and phospholipid. Involved in stabilization of microtubules.

Specifications

Antigen MID1IP1
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MID1IP1
Gene Accession No. Q6P7D5, Q9CQ20, Q9NPA3
Gene Alias G12 like, Mid1 interacting protein 1, MID1IP1, MIG12, Protein STRAIT11499, STRAIT11499, THRSPL
Gene Symbols Mid1ip1
Host Species Rabbit
Immunogen MID1IP1 Fusion Protein Ag8400
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404280, 58526, 68041
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.