man
Test
Search
Order Status
Quick Order
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Vials
Sample Vials
15118112
Print
Kinesis™ Microwave Vials
Brand:
Kinesis™ 98998-93
Additional Details :
Weight :
0.50000kg
Product Code.
15118112
4760.00 NOK
/ Pack of 50
Quantity
Estimated Shipment: 01-07-2024
Log in
Description
KX, for use with Initiator Systems (EXP only) from Biotage, contamination free 1st class hydrolytic borosilicate glass, resealable septa, 40°C to 300°C, 10mL to 20mL