Description
For intense mixing, dispersing, homogenizing, emulsifying and mincing operations for cuttable materials
- Mixing, dispersing, homogenizing, emulsifying with liquids
- Dust-free processing of dry samples such as hay
Specifications
Specifications
| Product Type | Laboratory Mixer |
| Speed | 1000 to 15,000 rpm |
| Voltage | 230 V |
| Wattage | 550 W |
| Volume Range (Metric) | 125 mL to 1 L |
| Height (Metric) | 245 mm |
| For Use With (Application) | For mixing, dispersing, homogenising, emulsifying and size reduction of cuttable materials |
| Model | MB 550 with EU Power Cable |
| Noise Level | 72 dBA at 15 000 rpm |
| Weight (Metric) | 4.4 kg |