Learn More
Thermo Scientific™ Microtiter Tubes and Racks
Shop All MBP Molecular BioProducts Products
Click to view available options
Product Type:
Frosted MicroTiter Tube
Microtiter Tube
Sterility:
Non-sterile
Sterile
Format:
Bulk
Robotic Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 960
Description
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
- Polyethylene plug strip seals the microtiter tubes tightly for storage
- Eight-plug strips can be easily cut apart for individual samples
- 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
- Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
- Microtiter tube system is available in bulk packaging or with a SBS standard footprint
- Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
- Rigorous testing to ensure sample integrity and security
Packaging Options
- Microtiter tubes and caps are available sterile and nonsterile
Spezifikation
Spezifikation
| Certifications/Compliance | Certified free of RNase, DNase, DNA and pyrogens |
| Color | Natural |
| Volume (Metric) | 1.2 mL |
| Quantity | 96 tubes/tray 10 trays/pack, nonsterile |
| Closure Material | Polyethylene (Plug) |
| Product Line | Thermo Scientific |
| Sterility | Non-sterile |
| Format | Robotic Rack |
| Product Type | Microtiter Tube |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur