Learn More
Greiner Bio-One Microscopic Cover slip for Terasaki Plates
Product Code. 15985311
Change view
Click to view available options
Microscopic coverslip
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15985311 Microscopic coverslip
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15985311 Supplier Greiner Bio-One Supplier No. 653081
  • ONLINE EXCLUSIVE PRICE
    66203.79 NOK
    57110.00 NOK / Pack of 5000
    Save 9093.79 NOK
    13% Off
Estimated Shipment: 08-10-2026
For use with Terasaki plates. Greiner Bio-One Microscopic Cover slip is manufactured from crystal clear polystyrene.
  • Manufactured from crystal clear polystyrene
  • Non-sterile
TRUSTED_SUSTAINABILITY
Specifications
Material Polystyrene
Length (Metric) 76 mm
Width (Metric) 51 mm
For Use With (Equipment) Terasaki Plates