Learn More
Heathrow Scientific™ Microscope Slide Holder
Product Code. 15346167 Shop All Thermo Scientific CO₂ Incubators Products
Product Code. 15346167 Supplier Heathrow Scientific™ Supplier No. HS15981C
  • ONLINE EXCLUSIVE PRICE
    447.00 NOK
    386.03 NOK / Pack of 1
    Save 60.97 NOK
    13% Off
View all promotions
 Restricted Item
Estimated Shipment: 12-01-2027
Holds slides during evaluation, cleaning, and drying. Heathrow Scientific™ includes rubber feet for to prevent unintentional movement.
  • Made from chemical-resistant polyethylene
  • Solid, strong construction with rubber feet for bench top stability
Specifications
Type Microscope Slide Box
Slide Capacity 15
Device Holds Slides
Dimensions (L x W x H) 153 x 102 x 21 mm
For Use With (Equipment) Microscope Slide
Material Polyethylene