Learn More
Fisherbrand™ Achromatic Microscope Objective
Product Code. 12308518
Click to view available options
Magnification Power:
60X
Unit Size:
Each
Product Code. 12308518
Brand: Fisherbrand™ 1220.0606
Use this objective with compound biological microscopes. Fisherbrand™ Achromatic Microscope Objectives are ergonomically designed for everyday use.
  • Achromatic microscope objective
  • Accessory for microscope models: FB69019 and FB69020
Specifications
Type Achromat Objective
Description 60x/0.85 (dry)
For Use With (Equipment) Microscope models: FB69019 and FB69020
Magnification Power 60X