Learn More
Fisherbrand™ Plan Achromatic Microscope Objective PH+
Product Code. 12338608
Click to view available options
Magnification Power:
25X
Unit Size:
Each
Product Code. 12338608
Brand: Fisherbrand™ 3623.0259
Correct for color while using an inverted microscope.
  • For use with inverted microscopes
  • Corrects for color
Specifications
Type Plan Achromatic Objective
Description 25x/0.40/160/1.2 LWD PH+
For Use With (Equipment) Microscope model: FB69196
Magnification Power 25X