missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microscope Eyepiece with Cross Line Scale

Product Code. 11522673
Click to view available options
Magnification Power:
10X
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11522673

Brand: Fisherbrand™ 1713.1001

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use this eyepiece with compound microscopes. Fisherbrand™ Microscope Eyepieces with Cross Line Scales are ergonomically designed for everyday use.

  • For Binocular Stereo Microscope FB69155

Specifications

Type Microscope Eyepiece with Cross Line Scale, 0 to 14 mm
Diameter (Metric) 18 mm
Magnification Power 10X
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.