Learn More
Buerkle™ V4A MicroSampler Head
Click to view available options
Capacity (Metric):
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
For Use With (Equipment):
12 mm dia. MicroSampler Tube
25 mm dia. MicroSampler Tube
Unit Size:
Each
Description
- Interchangeable
- Allows tips with different volumes to be used
- Stainless steel AISI 316L
Specifications
Specifications
| Product Type | MicroSampler Head |
| Material | Stainless Steel (V4A) |
| Autoclavable | Autoclavable |
| Description | Head for MicroSampler |
| Capacity (Metric) | 10 mL |
| For Use With (Application) | For The Sampling of Small Quantities of Powder |
| For Use With (Equipment) | 25 mm dia. MicroSampler Tube |