Learn More
Micronova™ NovaWipe™ Wipers
Product Code. 13177326 Shop All Micronova Products
Change view
Click to view available options
Dimensions (L x W):
12 x 10 in. (30.48 x 25.4 cm)
12 x 12 in.
18 x 4 in. (45.7 x 10.2 cm)
9 x 7 in. (22.9 x 17.8 cm)
9 x 9 in. (22.86 x 22.86 cm)
Packaging Quantity:
1 Ea.
10 Wipers/Pk.
Unit Size:
Pack of 100
Pack of 50
Product Code. Dimensions (L x W) Packaging Quantity unitSize
13177326 12 x 10 in. (30.48 x 25.4 cm) 10 Wipers/Pk. Pack of 100
13127336 12 x 12 in. 10 Wipers/Pk. Pack of 100
13137336 18 x 4 in. (45.7 x 10.2 cm) 10 Wipers/Pk. Pack of 50
13177336 9 x 7 in. (22.9 x 17.8 cm) 10 Wipers/Pk. Pack of 100
13187336 9 x 9 in. (22.86 x 22.86 cm) 10 Wipers/Pk. Pack of 100
13147336 9 x 9 in. (22.86 x 22.86 cm) 1 Ea. Pack of 100
6 options
Product Code. 13177326
Brand: Micronova™ WP11012
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For semiconductor and microelectronics cleaning where the fast-drying, absorbent properties of PVA are key
  • With a 600% absorbency, they are soft and pliable for both wet and dry applications
  • For general wiping, lapping and polishing applications
  • Can be laundered and re-used
Specifications
Product Type NovaWipe™ Cleanroom Wipes
Length (English) 12 in.
Length (Metric) 30.5 cm
Width (English) 10 in.
Width (Metric) 25.4 cm
Material Polyvinyl Alcohol
Disposable Reusable
Dimensions (L x W) 12 x 10 in. (30.48 x 25.4 cm)
For Use With (Application) Ideal for general wiping, lapping and polishing applications
For Use With (Equipment) Workstations, laminar flow benches, walls, windows
Packaging Quantity 10 Wipers/Pk.