Learn More
Hettich Lab Technology™ Microlitre Centrifuge
Product Code. 10792314
Change view
Click to view available options
Capacity:
30 x 1.5/2 mL
60 x 1.5/2 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
10792314 60 x 1.5/2 mL Each
10274042 30 x 1.5/2 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10792314 Supplier Hettich Lab Technology™ Supplier No. 2205
 Restricted Item
Estimated Shipment: 01-12-2026
The Microlitre Centrifuge can process very small samples at very high speeds. The powerful, maintenance free motors achieve high RCF values for best sample results. Reliable and steady temperature control, quick pre-cool and stand-by cooling functions maintain a constant sample temperature.
  • Better results and more samples in less time - increase efficiency and throughput in your lab
  • Wide range of capabilities due to the variety of models and accessories available
  • Pre-cooling function for fast operation - reliable and steady temperature control systems with quick pre-cool and standby functions for precise temperature control
Specifications
Capacity 60 x 1.5/2 mL
Max. Speed 18,000 rpm
Max. RCF 31,514 x g
Includes Angle Rotor
Safety Features Lid dropping protection
Depth (Metric) 650 mm
Product Line MIKRO 220R
Product Type Microlitre Centrifuge
Temperature Range (Metric) -20°C to +40°C