Learn More
Hamilton™ Microliter™ and Gastight™ Syringes: Model 701 for Agilent 7673, 7683, 7693 and 6850 ALS GC
Product Code. 10340281
Change view
Click to view available options
Needle Gauge:
23 to 26s G
23s G
26s G
Model:
701 N (Cemented Needle)
701 RN (Removable Needle)
Needle Point Number:
2
AS
Quantity:
6 Pk.
Ea.
Unit Size:
Each
Pack of 6
Product Code. Needle Gauge Model Needle Point Number Quantity unitSize
10340281 23 to 26s G 701 N (Cemented Needle) AS Ea. Each
10722711 23s G 701 N (Cemented Needle) AS 6 Pk. Pack of 6
10508502 23s G 701 RN (Removable Needle) AS Ea. Each
11567614 26s G 701 N (Cemented Needle) 2 Ea. Each
10376741 23s G 701 N (Cemented Needle) AS Ea. Each
10300331 23 to 26s G 701 RN (Removable Needle) AS Ea. Each
10117070 23 to 26s G 701 N (Cemented Needle) AS 6 Pk. Pack of 6
10156970 26s G 701 N (Cemented Needle) AS Ea. Each
10442721 26s G 701 RN (Removable Needle) AS Ea. Each
10347012 26s G 701 N (Cemented Needle) AS 6 Pk. Pack of 6
10055630 23s G 701 N (Cemented Needle) 2 Ea. Each
11 options
Product Code. 10340281
Brand: Hamilton™ 80393
Designed to work with Agilent 7673, 7683, 7693 and 6850 Automatic Liquid Sampler (ALS) gas chromatography (GC) autosamplers
Microliter syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling.
Syringe Terminations
  • Cemented Needle (N) - For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
  • Removable Needle (RN) - The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Styles
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 23s or 26s
  • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 23s, 26s, 23s-26s
Specifications
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Needle Gauge 23 to 26s G
Diameter (English) Inner 0.019 in.
Diameter (English) Inner Needle 0.0133 to 0.005 in.
Diameter (Metric) Inner Needle 0.64 to 0.13 mm
Diameter (English) Outer Needle 0.0283 to 0.0187 in.
Diameter (English) Outer 0.26 in.
Diameter (Metric) Inner 0.485 mm
Diameter (Metric) Outer 6.6 mm
Diameter (Metric) Outer Needle 0.72 to 0.47 mm
Length (English) 1.7 in.
Length (English) Needle 1.71 in.
Length (Metric) 43 mm
Product Type Microliter Syringe
Max. Pressure 2000 psi
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 701 N (Cemented Needle)
Needle Point Number AS
Min. Temperature (Metric) 10°C
Quantity Ea.
Volume (Metric) 10 μL
Needle Point Style Cone
For Use With (Equipment) Agilent 7673, 7683, 6850 Automatic Liquid Sampler, Gas Chromatography Autosamplers
Connection Type Cemented