Description
- Multi-layer barrier laminate
- Elasticated opening
- Fits over Euro sizes 42 to 46. (UK 8 to 11)
Specifications
Specifications
| Size | Fits over Euro shoe sizes 42 to 46 (UK 8 to 11) |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Closure Type | Elastic with Adjustable Ties |
| Seam Type | Ultrasonically Welded |
| Certifications/Compliance (Europe) | Category I PPE according to EuropeanDirective 89/686/EEC |
| Packaging Quantity | 150 Pairs |
| Product Type | Boot Covers |