Learn More
Ansell Edmont™ Microgard™ 2000 Standard Sleeves
Product Code. 15516544 Shop All Ansell Products
Change view
Click to view available options
Length (English):
21 in.
Unit Size:
300 pairs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
15516544 21 in. 300 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15516544 Supplier Ansell Edmont™ Supplier No. WH20B0060000
  • ONLINE EXCLUSIVE PRICE
    10699.50 NOK
    7070.28 NOK / 300 pairs
    Save 3629.22 NOK
    33% Off
View all promotions
Estimated Shipment: 01-12-2026
Fits over the arm just past the elbow to the bicep. Ansell Edmont™ Microgard™ 2000 Standard Sleeves include elasticated openings at both ends.
  • Elasticated at both ends
  • Bound seams
  • Microporous polyethylene laminate
  • Barrier to hazardous liquid and particulate substances
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Microporous Polyethylene Laminate (Non-woven)
Color White
Sterility Non-sterile
Disposable Single-use
Closure Type Elastic Ends
Seam Type Bound
Cuff Style Elastic
Length (English) 21 in.
Length (Metric) 51 cm