Learn More
Ansell™ Microgard™ 1500 Plus FR Coveralls with Hood
Product Code. 30058133 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Each
Pack of 30
Pack of 40
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058133 5X-Large Pack of 30
15546574 Medium Pack of 40
15566574 X-Large Pack of 40
15556574 Large Pack of 40
15576574 2X-Large Pack of 30
15586574 3X-Large Pack of 30
17131035 Small Pack of 40
30058132 4X-Large Pack of 30
18020130 4X-Large Each
18050130 5X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 30058133 Supplier Ansell™ Supplier No. WR17S0011109
Estimated Shipment: 30-12-2026
MICROGARD™ 1500 Plus FR Coveralls with Hood are made of a highly breathable anti-static SMS fabric which utilises the latest developments in micro fibre technology to ensure good filtration efficiency.
  • Category III Coverall with 3-piece hood
  • Stitched seams (Red colour)
  • Elasticated wrists, finger loops, waist and ankles.
  • 2-way front zipper with re-sealable storm flap.
  • Flame retardant and anti-static
  • Nonwoven SMMS polypropylene
  • Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
  • Anti-staticaccording to EN1149-5
  • Optimized body fit ensures full freedom of movement when worn over heat and flame protective clothing (EN ISO 14116 Index 2 or above).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Flame-resistant Coveralls
Size 5X-Large
Gender Unisex
Material FR Anti-static SMS Fabric
Color White
Disposable Reusable
Waist Style Elastic at Back
Ankle Style Elastic
Cuff Style Elastic, Finger Loops
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class CE CATEGORY III (Type 5, Type 6, EN 1073-2, EN ISO 14116, EN 1149-5)
Resistance Chemicals, Anti-static, Flame
Collar Type 3-piece elasticated hood
Certifications/Compliance (Europe) CE 0598, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, Beperkte vlamverspreiding EN ISO 14116: 2015-1 Index 1
Latex Free Yes
For Use With (Application) Chemical Handling, Contaminated Environments, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletization