Learn More
Cytiva (formerly Pall Lab) MicroFunnel™ Filters Funnel with Metricel™ Black Membrane
Ready-to-use, sterile and individually packaged MicroFunnel filter funnels reduce chances of cross contamination.
Brand:  Cytiva (formerly Pall Lab) 4805
Additional Details : Weight : 2.13750kg
 View more versions of this product
Product Code. 17104411
  • 4025.00 NOK / Pack of 50
Estimated Shipment: 28-11-2023
Description
  • Easy squeeze removal of funnel to access membrane – no twisting means less RSI and membrane tearing
  • Proven membrane performance
  • For research use only
Specifications
Specifications
Sterile
0.45 μm
50/Pk.
Metricel Membrane Modified PES
Black
100 mL