Description
- Apron with Sleeves
- Rear velcro fastening
- Elasticated wrists
- Welded seams
- Multi-layer barrier fabric effective against numerous chemicals
- Highly visible bright yellow for improved worker safety
- Lightweight yet durable
- Anti-static according to EN 1149-5
Specifications
Specifications
| Type | Protective Apron |
| Size | X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Closure Type | Hook and Loop (Rear) |
| Cuff Style | Elastic |
| No. of Pockets | None |
| Certifications/Compliance | Type PB 3/4 EN 14605, Type PB(3)-B, PB(4)-B EN 14126, EN1149-1 |
| Seam Type | Ultrasonically Welded |