Learn More
Cytiva Microcheck™ Beverage Monitor
Product Code. 17124401
Change view
Click to view available options
Material (Membrane):
GN-6 Metricel Mixed Cellulose Ester
Metricel Black Membrane Modified PES
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Membrane) unitSize
17124401 Metricel Black Membrane Modified PES Pack of 50
17104401 GN-6 Metricel Mixed Cellulose Ester Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17124401 Supplier Cytiva Supplier No. 4763
  • ONLINE EXCLUSIVE PRICE
    1985.00 NOK
    1849.71 NOK / Pack of 50
    Save 135.29 NOK
    6% Off
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Easy to use, versatile, bulk packaged disposable filter funnels are ideal for microbial analysis.
  • Can convert to a petri dish for culturing or easily remove the membrane for placing on separate agar dish
  • Product not approved for use in medical clinical surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 100 mL
Color Black
Description Polystyrene Cover, Polypropylene Cylinder/Base, Polyethylene Adapter Material, Cellulose Support Pad
Pore Size 0.45 μm
Quantity 50/Pk.
Sterility Gamma-irradiated
Material (Membrane) Metricel Black Membrane Modified PES