Learn More
Ismatec™ Platinum-Cured Silicone Microbore 2-Stop Tube Sets
Click to view available options
Diameter (Metric) Inner:
2.06 mm
2.79 mm
Diameter (Metric) Outer:
3.76 mm
4.49 mm
Flow Rate:
130 mL/min.
200 mL/min.
Unit Size:
Pack of 6
Description
- Compatible with Masterflex L/S Multichannel pump head, small cartridges
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most formulations and sizes available to ship same day
Specifications
Specifications
| Description | Microbore 2-Stop tube sets, silicone (platinum-cured); 2.79 mm ID. Pk of 6. |
| Diameter (English) Inner | 0.11 in. |
| Diameter (Metric) Inner | 2.79 mm |
| Diameter (English) Outer | 0.176 in. |
| Diameter (Metric) Outer | 4.49 mm |
| Overall Length (English) | 1.25 ft. |
| Overall Length (Metric) | 0.38 m |
| Tubing Size | 48 |
| Flow Rate | 200 mL/min. |
| Hose Barb | 3/32 in. Tubing |
| Show More |