Learn More
Ismatec™ Platinum-Cured Silicone Microbore 2-Stop Tube Sets
Product Code. 11705159
Click to view available options
Diameter (Metric) Inner:
2.06 mm
2.79 mm
Diameter (Metric) Outer:
3.76 mm
4.49 mm
Flow Rate:
130 mL/min.
200 mL/min.
Unit Size:
Pack of 6
Product Code. 11705159
Brand: Ismatec™ 0642142
Only null left
Add to basket
Add to basket
Choose the size and formulation to suit your flow rate and application needs.
  • Compatible with Masterflex L/S Multichannel pump head, small cartridges
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most formulations and sizes available to ship same day
Specifications
Description Microbore 2-Stop tube sets, silicone (platinum-cured); 2.06 mm ID. Pk of 6.
Diameter (English) Inner 0.081 in.
Diameter (Metric) Inner 2.06 mm
Diameter (English) Outer 0.147 in.
Diameter (Metric) Outer 3.76 mm
Overall Length (English) 1.25 ft.
Overall Length (Metric) 0.38 m
Tubing Size 42
Flow Rate 130 mL/min.
Hose Barb 3/32 in. Tubing
Maximum Pressure 25.3 psi