Learn More
Applied Biosystems™ MicroAmp™ Optical 96-Well GPLE Reaction Plates with Barcode
Product Code. 15266545 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15266545 10 Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15266545 Supplier Applied Biosystems™ Supplier No. 4481192
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Heat transfer from thermal block to sample with thin-wall polypropylene wells
  • Applied Biosystems MicroAmp Optical 96-Well GPLE Reaction Plates with Barcode are validated and tested for optimal fit and performance on Applied Biosystems standard (0.2 mL) 96-well thermal cyclers, 0.2-mL real-time PCR instruments, and genetic analyzers.
  • Features of the MicroAmp Optical 96-Well GPLE Reaction Plates include:
  • Each lot is certified DNA-, RNase-, and PCR inhibitor-free
  • Labeled “For Laboratory Use”
  • Optimized for performance with Applied Biosystems instruments
  • Excellent heat transfer from thermal block to sample with thin-wall polypropylene wells
  • Raised deck for efficient plate sealing to help prevent evaporation and minimize contamination
  • MicroAmp Optical 96-well reaction plates are constructed from a single rigid piece of polypropylene and include a raised deck for efficient plate sealing.
  • The thin-wall polypropylene wells provide excellent heat transfer from the thermal block to the sample for fast, efficient PCR amplification.
  • These MicroAmp Optical 96-well reaction plates contain a single barcode (including a human readable barcode) on the south (row H1-H12) side of the plate.
  • With General Purpose Lab Equipment (GPLE) plastics, each lot of reaction plates is certified in an ISO 13485-registered facility to be free of DNA, RNase, and PCR inhibitors. GPLE plastics are ideal for use in diagnostic procedures.
TRUSTED_SUSTAINABILITY
Specifications
Barcode Includes Barcode
Certifications/Compliance Certified DNA/RNase Free
Packaging 10 Plates
Product Line MicroAmp
Color Optical
Product Type Microplate
For Use With (Equipment) Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt Style Semi
Material Polypropylene
No. of Wells 96
Volume (Metric) Working 0.2 mL
Quantity 10 Plates
Shipping Condition Room Temperature