Learn More
Ansell MICRO-TOUCH™ Nitrile Gloves
Product Code. 17803642 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17803642 Large Pack of 100
17863702 X-Small Pack of 100
17843802 Small Pack of 100
17823802 Medium Pack of 100
17803752 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17803642 Supplier Ansell Supplier No. 313029090
  • ONLINE EXCLUSIVE PRICE
    147.00 NOK
    117.09 NOK / Pack of 100
    Save 29.91 NOK
    20% Off
View all promotions
Estimated Shipment: 23-11-2026
MICRO-TOUCH™ Nitrile Gloves feature a thin, yet durable nitrile construction to provide healthcare professionals the reliable protection and tactility they need. These gloves are approved for chemotherapy use.
  • Designed with a thin, yet durable nitrile formulation for dependable protection
  • Royal blue color provides greater contrast than standard light blue nitrile gloves
  • Approved for use with chemotherapy drugs
Specifications
Applications Healthcare
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 5.1 mil
Finger Thickness (Metric) 0.13 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Large
Quantity 100 Gloves/Box, 10 Boxes/Cs.