Learn More
Ansell MICRO-TOUCH Affinity Gloves
Product Code. p-200080401 Shop All Ansell Products
Click to view available options
Size:
Large, 8.5/9
Medium, 7.5/8
Small, 6.5/7
X-Large, 9.5/10
Unit Size:
Pack of 100
Product Code. 16306901
Brand: Ansell 553352
Only null left
Add to basket
Add to basket
MICRO-TOUCH Affinity Gloves are neoprene, powder-free examination gloves with excellent elasticity and barrier protection.
  • Robust Neoprene formulation provides comfort and excellent tear and puncture resistance
  • Textured fingers for secure grip
  • Protects from Type I latex allergy
  • Tested for use with chemotherapy drugs
Specifications
Accelerator Free Yes
Applications Laboratory
Color Green
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Polychloroprene
Powdered Powder Free
Product Type Exam Gloves
Size Small, 6.5/7