Learn More
Kartell SpA Micro Test Tube
Description
Allows for centrifugation up to RCF 21,000 x g. Large opening tab featuring rounded surfaces for smoother open/close routine and cap top with 3 mm pierceable area. Autoclavable at 121°C for 20 minutes with open cap.
Specifications
Specifications
| Cap Style | Flat cap |
| Certifications/Compliance | CE/IVD |
| Closure Color | Pink |
| Closure Material | PP (Polypropylene) |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 1.5 mL |
| Color | Pink |
| Material | Polypropylene |
| For Use With (Application) | Sample analysis |
| No. per Pack | 1000/Pk. |
| Show More |