Learn More
Wheaton™ Micro-Stir™ Single Place Magnetic Stirrer
Product Code. 10354271 Shop All Wheaton Science Products Products
Click to view available options
Plug Type:
EU
Unit Size:
Each
Product Code. 10354271
Brand: Wheaton™ W900700C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured specifically for cell culture applications. Wheaton™ Micro-Stir™ Single Place Magnetic Stirrer is ideal for low speed shear mixing.
  • 5-200 RPM speed range
  • Programmable, remote control and alarm capable
  • Brushless direct drive motor provides low maintenance 
TRUSTED_SUSTAINABILITY
Specifications
Speed 5 to 200 rpm
Stirring Positions 1
Top Plate Material Aluminum
Voltage 230 V
Wattage 15 w
Height (Metric) 92 mm
Width (Metric) 203 mm
Electrical Requirements 100 to 240V, 50/60 Hz, 15w
Frequency 50/60 Hz
Power Setting Programmable, remote control
Plug Type EU
Temperature Range (Metric) 15°C to 40°C
Type Magnetic Stirrer
Weight (Metric) 1.9 kg