Learn More
SI Analytics MICRO-OSTWALD STD 40 - 800mm²/S
Brand:  SI Analytics 285404441
MICRO-OSTWALD STD 40 - 800mm²/S
Product Code. 10161401
  • ONLINE EXCLUSIVE PRICE
    3065.00 NOK
    2725.82 NOK / Each
    Save 339.18 NOK
    11% Off
Only null left
Estimated Shipment: 23-01-2026