Learn More
ROBU VitraPOR™ Micro-Filter Candle with Glass Tube
Micro-Filter Candle with narrow glass tube.
Brand:  ROBU VitraPOR™ 18120
Additional Details : Weight : 0.11540kg
 View more versions of this product
Product Code. 16892936
  • 2220.00 NOK / Pack of 5
Estimated Shipment: 29-05-2024
Description
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Specifications
Borosilicate Glass (3.3)
Glass Tube
25 mm
Non-sterile
0
13 mm