missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Monoclonal Antibody (OTI4C8), TrueMAB™, OriGene
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | MICB |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI4C8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | MICB |
| Gene Accession No. | Q29980 |
| Gene Alias | MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC I like leukocyte 2; MICB; MIC-B; Mill2; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; PERB11.2; sMICB; soluble MICB; stress inducible class I homolog |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?