missing translation for 'onlineSavingsMsg'
Learn More

MI-136, MedChemExpress

Product Code. 16453697
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16453697

Brand: Medchem Express HY19319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors.

MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors.

Specifications

Chemical Name or Material MI-136
CAS 1628316-74-4
Color White
Purity Grade Notes Research
Molecular Formula C23H21F3N6S
Quantity 50 mg
Solubility Information DMSO : 110 mg/mL (233.79 mM; Need ultrasonic)
SMILES FC(F)(F)CC(S1)=CC2=C1N=CN=C2NC3CCN(CC4=CC=C(NC(C#N)=C5)C5=C4)CC3
Molecular Weight (g/mol) 470.51
Formula Weight 470.51
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.