Learn More
MHP, MedChemExpress
Product Code. 16460779
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
Product Code. Quantity unitSize
16460779 1 mg 1mg
16443428 10 mg 10mg
16430416 20 mg 20mg
16406157 5 mg 5mg
4 options
Product Code. 16460779
Brand: Medchem Express HY101572
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress MHP (Methyl caprooyl tyrosinate) is an activator of sphingosine kinase (SPHK1), and significantly stimulates CAMP mRNA and protein production. MHP (Methyl caprooyl tyrosinate) enhances antimicrobial defense and innate immunity.
MHP (Methyl caprooyl tyrosinate) is an activator of sphingosine kinase (SPHK1), and significantly stimulates CAMP mRNA and protein production. MHP (Methyl caprooyl tyrosinate) enhances antimicrobial defense and innate immunity.
Specifications
Chemical Name or Material MHP
CAS 1104874-94-3
Color White
Purity Grade Notes Research
Molecular Formula C16H23NO4
Quantity 1 mg
Synonym Methyl caprooyl tyrosinate
Solubility Information DMSO : ≥ 100 mg/mL (340.88 mM) ∣H2O : 1 mg/mL (3.41 mM; Need ultrasonic)
SMILES OC(C=C1)=CC=C1C[C@@H](C(OC)=O)NC(CCCCC)=O
Molecular Weight (g/mol) 293.36
Formula Weight 293.36
Percent Purity 98.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation