missing translation for 'onlineSavingsMsg'
Learn More

Mhc Rabbit anti-Drosophila, Polyclonal, Proteintech

Product Code. p-7088276 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16857423 20 μL 20µL
16847423 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16857423

Brand: Proteintech 107991AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects the Myosin heavy chain of drosophila.

The function of Mhc is muscle contraction.

Specifications

Antigen Mhc
Applications ELISA
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene mhc
Gene Accession No. P05661
Gene Alias Bsh, CG17927, Dm II, Dmel\CG17927, DmMHC, DROMHC, DroMII, FBgn0002741, Ifm(2)2, l(2)36Ae, l(2)k10423, l(2)M66, Mhc, Mhc36B, mMHC, MRP, Myo, Myosin heavy chain, Nup, Sht, sMHC, Stp
Gene Symbols mhc
Host Species Rabbit
Immunogen Mhc Fusion Protein Ag1254
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 35007
Target Species Drosophila
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.