missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MHC Class I (H-2Kd) Monoclonal Antibody (SF1-1.1.1), Super Bright™ 600, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
1544.00 NOK - 3470.00 NOK
Specifications
| Antigen | MHC Class I (H-2Kd) |
|---|---|
| Clone | SF1-1.1.1 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16321524
|
Invitrogen™
63-5957-80 |
25 μg |
1544.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16331524
![]() |
Invitrogen™
63-5957-82 |
100 μg |
3470.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC Class I (H-2Kd) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MGC184092; MGC7052; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
| H2-K1 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| H2-K1 |
| SF1-1.1.1 | |
| Flow Cytometry | |
| Super Bright 600 | |
| Mouse | |
| Mouse | |
| P01901, P03991, P04223, P14428 | |
| 14972 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title