Learn More
Invitrogen™ MHC Class I (H-2Kd) Monoclonal Antibody (SF1-1.1.1), Biotin, eBioscience™

Mouse Monoclonal Antibody
1492.00 NOK
Specifications
| Antigen | MHC Class I (H-2Kd) |
|---|---|
| Clone | SF1-1.1.1 |
| Concentration | 0.5 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Flow Cytometry |
Description
Description: This SF1-1.1.1 monoclonal antibody reacts with the H-2Kd MHC class I alloantigen. H-2Kd is involved in antigen presentation to T cells expressing CD3/TCR and CD8. Reactivity to other haplotypes (e.g. b, j, k, p, q, s, and v) has not been observed. Applications Reported: This SF1-1.1.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SF1-1.1.1 antibody has been tested by flow cytometric analysis of Balb/c splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC Class I (H-2Kd) | |
| 0.5 mg/mL | |
| Flow Cytometry | |
| Biotin | |
| Mouse | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P01901, P03991, P04223, P14428 | |
| 14972 | |
| Primary | |
| Affinity chromatography |
| SF1-1.1.1 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Monoclonal | |
| Liquid | |
| IgG2a κ | |
| Mouse | |
| H2-K1 | |
| class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K; H2-K; H-2K(B); H-2K(d); H-2K(K); H-2K(Q); H2-K1; histocompatibility 2, K1, K region; K-f; MGC184092; MGC7052; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen | |
| H2-K1 | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.