missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I Antibody (OX18) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB120-6405SS

1661.75 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


 View more versions of this product

Product Code. 18515372

  • 2125.00 NOK / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MHC Class I Monoclonal antibody specifically detects MHC Class I in Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, CyTOF, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MHC Class I
Monoclonal
1 mg/mL
Flow Cytometry 1:50-1:100, ELISA 1:100-1:2000. Use reported in scientific literature (PMID 2783579), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500. Use reported in scientific literature (PMID 24678820), Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Electron Microscopy reported in scientific literature (PMID 3044648), Flow (Intracellular), CyTOF-ready, Block/Neutralize reported in scientific literature (PMID 1698855)
HLA-E
Rat spleen glycoproteins
0.025 mg
Adaptive Immunity, Immunology, Major Histocompatibility Complexes
3133
Rat
Purified
Flow Cytometry
OX18
Unconjugated
Clone OX18, DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC Class I RT1-A, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, OX18, QA1, RT1-A, RT1-A (OX18)
Mouse
Protein G purified
RUO
Primary
Recognizes a monomorphic determinant of rat MHC Class I (RT1A), expressed by all rat strains. However, quantitative measurements suggest that not all of the class I molecules are recognised.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only