missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MGMT (Human) Recombinant Protein
Human MGMT full-length ORF ( AAH00824, 1 a.a. - 207 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004255-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MDKDCEMKRTTLDSPLGKLELSGCEQGLHEIKLLGKGTSAADAVEVPAPAAVLGGPEPLMQCTAWLNAYFHQPEAIEEFPVPALHHPVFQQESFTRQVLWKLLKVVKFGEVISYQQLAALAGNPKAARAVGGAMRGNPVPILIPCHRVVCSSGAVGNYSGGLAVKEWLLAHEGHRLGKPGLGGSSGLAGAWLKGAGATSGSPPAGRN
Specifications
AAH00824 | |
O-6-methylguanine-DNA methyltransferase | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
4255 | |
Wheat germ expression system | |
25 μg | |
MGMT | |
GST |