missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGAT5B Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | MGAT5B |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16856804
|
Proteintech
16993-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16846804
|
Proteintech
16993-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycosyltransferase that acts on alpha-linked mannose of N-glycans and O-mannosyl glycans. Catalyzes the transfer of N-acetylglucosamine (GlcNAc) to the beta 1-6 linkage of the mannose residue of GlcNAcbeta1,2-Manalpha on both the alpha1,3- and alpha1,6-linked mannose arms in the core structure of N-glycan. Also acts on the GlcNAcbeta1,2-Manalpha1-Ser/Thr moiety, forming a 2,6-branched structure in brain O-mannosyl glycan. Plays an active role in modulating integrin and laminin-dependent adhesion and migration of neuronal cells via its activity in the O-mannosyl glycan pathway.Specifications
| MGAT5B | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| Q3V5L5 | |
| 146664, 303693 | |
| MGAT5B Fusion Protein Ag10678 | |
| Primary | |
| -20°C | |
| MGAT5B |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GlcNAc T Vb, GnT IX, GnT VB, hGnTVb, KIAA2008, MGAT5B | |
| MGAT5B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title