missing translation for 'onlineSavingsMsg'
Learn More

MGAT2-IN-1, MedChemExpress

Product Code. p-200100775
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496337 5 mg 5mg
16470007 50 mg 50mg
16450929 1 mg 1mg
16434706 100 mg 100mg
16433618 10 mg 10mg
16412796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16496337 Supplier Medchem Express Supplier No. HY101857
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MGAT2-IN-1 is an orally active inhibitor of monoacylglycerol acyltransferase (MGAT2) with IC50 of 7.8 and 2.4 nM for human and mouse MGAT2, respectively.

MGAT2-IN-1 is an orally active inhibitor of monoacylglycerol acyltransferase (MGAT2) with IC50 of 7.8 and 2.4 nM for human and mouse MGAT2, respectively.

Specifications

Chemical Name or Material MGAT2-IN-1
CAS 1800025-30-2
Color Gray
Purity Grade Notes Research
Molecular Formula C27H21ClF5N7O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (191.13 mM)
SMILES O=S(C1=CC2=C(N(C3=NC=C(C4=CC(C(F)(F)F)=NN4C)C=N3)CC2)C(N5C(CCC5)=O)=C1)(NC6=C(F)C=C(Cl)C=C6F)=O
Molecular Weight (g/mol) 654.01
Formula Weight 654.01
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.