Learn More
MGAT2-IN-1, MedChemExpress
Product Code. 16496337
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496337 5 mg 5mg
16470007 50 mg 50mg
16450929 1 mg 1mg
16434706 100 mg 100mg
16433618 10 mg 10mg
16412796 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16496337 Supplier Medchem Express Supplier No. HY101857
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress MGAT2-IN-1 is an orally active inhibitor of monoacylglycerol acyltransferase (MGAT2) with IC50 of 7.8 and 2.4 nM for human and mouse MGAT2, respectively.
MGAT2-IN-1 is an orally active inhibitor of monoacylglycerol acyltransferase (MGAT2) with IC50 of 7.8 and 2.4 nM for human and mouse MGAT2, respectively.
Specifications
Chemical Name or Material MGAT2-IN-1
CAS 1800025-30-2
Color Gray
Purity Grade Notes Research
Molecular Formula C27H21ClF5N7O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 125 mg/mL (191.13 mM)
SMILES O=S(C1=CC2=C(N(C3=NC=C(C4=CC(C(F)(F)F)=NN4C)C=N3)CC2)C(N5C(CCC5)=O)=C1)(NC6=C(F)C=C(Cl)C=C6F)=O
Molecular Weight (g/mol) 654.01
Formula Weight 654.01
Percent Purity 99.49%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism