Learn More
MFN2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28341-1-AP-20UL
 View more versions of this product
Product Code. 16886095
  • 1710.33 NOK / 20µL
Description
The MFN2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MFN2. This antibody recognizes human, mouse, rat antigen. The MFN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MFN2
Polyclonal
Unconjugated
20 μL
9927
IgG
Western Blot, Immunohistochemistry, ELISA
467 μg/mL
Rabbit
Primary
Human, Rat, Mouse