Learn More
Invitrogen™ MFN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120367
Description
Positive test controls include: PC-3, Mouse kidney, Rat kidney. The target is usually found in the following locations: Cytoplasm, Mitochondrion outer membrane, Multi-pass membrane protein. Immunogen sequence: MAETVSPLKH FVLAKKAITA IFGQLLEFVT EGSHFVEATY RNPELDRIAS EDDLVEIQGY RNKLAVIGEV LSRRHMKVAF FGRTSSGKSS VINAMLWDKV.
Mitofusin 1 (MFN1) and the related protein MFN2 are mitochondrial membrane GTPase proteins that play a central role in mitochondrial metabolism and may be associated with obesity and/or apoptosis processes. MFN1 and MFN2 form homotypic and heterotypic complexes and coordinately regulate mitochondrial fusion and are essential for embryonic development. When ectopically expressed, MFN1 inhibits the apoptosis-associated amino-terminal conformation change in the apoptotic protein Bax but not its mitochondrial translocation, indicating that MFN1 is involved in the regulating the activation of Bax on the outer mitochondrial membrane.
Specifications
| MFN1 | |
| Polyclonal | |
| Unconjugated | |
| MFN1 | |
| 2310002F04Rik; 6330416C07Rik; D3Ertd265e; DKFZp762F247; EC 3.6.5; FLJ20693; Fzo homolog; Fzo1b; hfzo1; hfzo2; HR2; Mfn1; MFN2; MGC41806; mitochondrial mitofusin 1; mitochondrial transmembrane GTPase Fzo-1; mitochondrial transmembrane GTPase FZO1B; mitochondrial transmembrane GTPase FZO-2; mitofusin 1; mitofusin 2; mitofusin-1; mKIAA4032; putative transmembrane GTPase; transmembrane GTPase MFN1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 192647, 55669, 67414 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q811U4, Q8IWA4, Q8R4Z9 | |
| MFN1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse MFN1 (NP_077162.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.