Learn More
Mettler Toledo™ Expansion Modules for SevenExcellence™ Meters
Product Code. 13450689 Shop All Mettler Toledo Products
Change view
Click to view available options
Type:
Blank Module
Conductivity Module
pH/Ion Module
pH/mV Module
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
13450689 Blank Module Each
13410689 pH/mV Module Each
13430689 Conductivity Module Each
13440689 pH/Ion Module Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 13450689 Supplier Mettler Toledo™ Supplier No. 30034475
Only null left
Add to basket
Add to basket
The modular design of SevenExcellence allows you to expand your instrument with additional measurement parameters for future needs at any time.
Just add the new expansion unit, switch on the instrument and the new expansion unit is automatically recognized and ready to use! SevenExcellence fits in total three expansion units in any composition.
TRUSTED_SUSTAINABILITY
Specifications
Type Blank Module