Learn More
METTLER TOLEDO™ S479 SevenExcellence™ pH/Conductivity/DO Benchtop Meter Kit
Shop All Mettler Toledo ProductsDescription
Includes
SevenExcellence pH/mV/Conductivity/DO triple channel meter (instrument with a pH/mV, conductivity and DO expansion unit); as well as an InLab™ ExpertPro-ISM (pH), InLab 731-ISM (conductivity) and InLab OptiOx (digital DO) sensor; pH buffers; conductivity standards; and zero-oxygen calibration tablets; guides to pH, conductivity, and DO measurement; uPlace™ electrode holder, semi-transparent cover, operating instructions, installation, LabX™ direct pH PC software, declaration of conformity and test certificate
S479 meter kit containing SevenExcellence meter, electrode, and accessories to perform pH, conductivity, dissolved oxygen, resistivity, salinity, TDS, BOD, Redox/ORP, and ISFET measurements
- The SevenExcellence can be expanded with additional modules to measure pH, conductivity, resistivity, ISE, salinity, TDS, Redox/ORP, and ISFET
- Easily create measurement and calibration shortcuts
- 7 inch color touch screen with intuitive menu navigation
- Compatible with a wide range of electrodes for any application
- Compatible with our ISM electrodes that capture and save calibration data which is then automatically recognized by the meter once connected
- Meter connects to electrodes via BNC; S7 to BNC cables available
- Export data to a printer, Excel file, or LIMS system
- Automation available for measurements and cleaning in high-throughput applications
- Manage user permissions and access to ensure data integrity
- Compatible with LabX software for advanced data management and 21CFR part 11 regulatory compliance
- Optional comprehensive qualification package available including IQ/OQ/PQ
- Preventative maintenance and service available through METTLER TOLEDO Service
Specifications
Specifications
| Description | SevenExcellence pH/Cond/DO meter S479-K |
| Channels | Multiple |
| Display Type | Color TFT |
| IP Rating | IP54 |
| Measurement Modes | pH, Conductivity, Dissolved Oxygen |
| Percent Saturation Range Polarographic DO | 0 to 600% |
| Range (mV) | -2000 to 2000 mV |
| Range (pH) | -2.000 to 20.000 |
| Range (Resistivity) | 0.01 to 100 MOhm-cm |
| Range (Salinity) | 0 to 80 ppt |
| Show More |