Description
The ErgoClips safer dosing method substantially reduces contamination risks, enables fast dosing processes and increases productivity.
Specifications
Specifications
| For Use With (Equipment) | Test Tubes, Centrifuge Tubes or Small Plastic Tube |
| Length (English) | 1.2 in. |
| Length (Metric) | 3.2 cm |
| Width (English) | 1.2 in. |
| Width (Metric) | 3.2 cm |
| Type | ErgoClip™ Small Basket |