Learn More
Mettler Toledo™ Balance Printer Accessories: Cables
Product Code. 11947951 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
Printer Pin Style 15P-9S, to AT, UMT2, MT5 and BD
Length (English):
58.8 in.
Length (Metric):
150 cm
Unit Size:
Each
Product Code. For Use With (Equipment) Length (English) Length (Metric) unitSize
11947951 Printer Pin Style 15P-9S, to AT, UMT2, MT5 and BD 58.8 in. 150 cm Each
1 options
Product Code. 11947951
Brand: Mettler Toledo™ 229029
Printer accessories for use with Mettler Toledo balances.
For use with Mettler Toledo balances.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Printer Pin Style 15P-9S, to AT, UMT2, MT5 and BD
Length (English) 58.8 in.
Length (Metric) 150 cm
Type Connection Cable