missing translation for 'onlineSavingsMsg'
Learn More

METTL5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7207853 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16856534 20 μL 20µL
16846534 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16856534

Brand: Proteintech 167911AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Probable methyltransferase.

Specifications

Antigen METTL5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene METTL5
Gene Accession No. Q8K1A0, Q9NRN9
Gene Alias HSPC133, methyltransferase like 5, METTL5
Gene Symbols METTL5
Host Species Rabbit
Immunogen METTL5 Fusion Protein Ag10232
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29081, 502632, 75422
Target Species Mouse, Rat, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.