Learn More
Metro™ MetroMax iQ™ Stainless-Steel Stem Casters
Description
- Promotes easy cleaning
- Casters allow shelving units to be moved easily for access to floors and walls
- When used to create carts, allows efficient movement of supplies throughout a facility or campus
Polymer Stem Casters
- Corrosion-resistant performance (all-polymer horn, stainless-steel axle and hardware)
- 5 in. (127mm) diameter polyurethane wheel
- Sleeve-stye axle bearing
- Optional toe-operated brake mechanism
- Antimicrobial product protection built into tread
- Load rating of 300 lbs. (135kg)
- Temperature range: -20 to 120°F (-29 to 49°C) [continuous]
- Bumpers included
Standard Stem Casters
- Plated caster horns and hardware
- Stem/Swivel or Stem/Brake style
- Bumpers included
Stainless-steel, Cart-Washable Stem Caster
- Stainless horn, axle, and stem
- Sealed bearings (bumper included)
Specifications
Specifications
| Product Type | Swivel Stem Caster |
| Diameter (English) | 5 in. |
| Diameter (Metric) | 12.7 cm |
| For Use With (Equipment) | Metro™ MetroMax i™ and Q™ shelving |
| Load Bearing Capacity (English) | 300 lb. |
| Load Bearing Capacity (Metric) | 135 kg |
| Temperature Range (English) | -20 to 120°F |
| Temperature Range (Metric) | -49 to 49°C |
| Wheel Tread | Polyurethane |
| With Casters | Yes |
| Show More |